siRNA Details
| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133537.2 | NP_598221.2 | 2 | 101 | ||
| GenBank mRNA | EF121998.1 | - | 2 | 47 | ||
| EF121999.1 | - | 2 | 47 | |||
| X13309.1 | - | 2 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc18| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133537.2 | NP_598221.2 | 2 | 101 | ||
| GenBank mRNA | EF121998.1 | - | 2 | 47 | ||
| EF121999.1 | - | 2 | 47 | |||
| X13309.1 | - | 2 | 90 | |||