siRNA Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133538.2 | NP_598222.2 | 10 | 1289 | ||
| XM_006257427.2 | XP_006257489.1 | 9 | 1157 | |||
| XM_006257428.2 | XP_006257490.1 | 9 | 1275 | |||
| XM_006257430.2 | XP_006257492.1 | 8 | 1088 | |||
| XM_006257431.2 | XP_006257493.1 | 6 | 796 | |||
| XM_008773388.1 | XP_008771610.1 | 9 | 1429 | |||
| XM_008773389.1 | XP_008771611.1 | 10 | 1316 | |||
| XM_008773390.1 | XP_008771612.1 | 6 | 890 | |||
| XM_008773391.1 | XP_008771613.1 | 6 | 1497 | |||
| GenBank mRNA | AF448818.1 | - | 10 | 1289 | ||

Il13ra2
Genomic Map