siRNA Details
| Gene Symbol: Mrpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133539.1 | NP_598223.1 | 3 | 325 | ||
| GenBank mRNA | BC058447.1 | - | 3 | 367 | 6919782 | |
| FQ212468.1 | - | 3 | 428 | |||
| FQ229319.1 | - | 3 | 380 | |||
| U53512.1 | - | 3 | 325 | |||

Mrpl17
Genomic Map