siRNA Details
| Gene Symbol: Gtf3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133541.1 | NP_598225.1 | 25 | 3996 | ||
| XM_006230205.2 | XP_006230267.1 | 25 | 4017 | |||
| XM_008759857.1 | XP_008758079.1 | 19 | 2967 | |||
| GenBank mRNA | L28801.1 | - | 25 | 3996 | ||

Gtf3c1
Genomic Map