siRNA Details
| Gene Symbol: Igsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133542.2 | NP_598226.1 | 2 | 389 | ||
| GenBank mRNA | AJ223184.1 | - | 2 | 389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf6| Gene Symbol: Igsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133542.2 | NP_598226.1 | 2 | 389 | ||
| GenBank mRNA | AJ223184.1 | - | 2 | 389 | ||