siRNA Details
| Gene Symbol: Pou2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100639.1 | NP_001094109.1 | 8 | 741 | ||
| XM_008769591.1 | XP_008767813.1 | 8 | 2867 | |||
| XM_008769592.1 | XP_008767814.1 | 8 | 1002 | |||
| XM_008769593.1 | XP_008767815.1 | 10 | 1201 | |||
| XM_008769594.1 | XP_008767816.1 | 9 | 1103 | |||
| XM_008769595.1 | XP_008767817.1 | 10 | 1136 | |||
| XM_008769596.1 | XP_008767818.1 | 10 | 3311 | |||
| XM_008769597.1 | XP_008767819.1 | 9 | 3213 | |||
| XM_008769598.1 | XP_008767820.1 | 8 | 3112 | |||
| XM_008769599.1 | XP_008767821.1 | 8 | 765 | |||
| GenBank mRNA | U17013.1 | - | 3 | 331 | ||

Pou2f1
Genomic Map