siRNA Details
Gene Symbol: Ppp1r15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133546.3 | NP_598230.2 | 2 | 811 | ||
XM_006228974.2 | XP_006229036.1 | 2 | 873 | |||
GenBank mRNA | AF020618.1 | - | 1 | 988 | ||
BC072513.1 | - | 2 | 793 | 7104357 |
