siRNA Details
| Gene Symbol: Csf2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133555.1 | NP_598239.1 | 13 | 2417 | ||
| GenBank mRNA | AJ000555.1 | - | 13 | 2417 | ||
| S79263.1 | - | 13 | 2417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf2rb| Gene Symbol: Csf2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133555.1 | NP_598239.1 | 13 | 2417 | ||
| GenBank mRNA | AJ000555.1 | - | 13 | 2417 | ||
| S79263.1 | - | 13 | 2417 | |||