siRNA Details
Gene Symbol: Csf2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133555.1 | NP_598239.1 | 13 | 2744 | ||
GenBank mRNA | AJ000555.1 | - | 13 | 2744 | ||
S79263.1 | - | 13 | 2744 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csf2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133555.1 | NP_598239.1 | 13 | 2744 | ||
GenBank mRNA | AJ000555.1 | - | 13 | 2744 | ||
S79263.1 | - | 13 | 2744 |