siRNA Details
| Gene Symbol: Itfg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133557.2 | NP_598241.2 | 8 | 869 | ||
| GenBank mRNA | AF480856.2 | - | 8 | 745 | ||
| BC085706.1 | - | 8 | 869 | 7189102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itfg1| Gene Symbol: Itfg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133557.2 | NP_598241.2 | 8 | 869 | ||
| GenBank mRNA | AF480856.2 | - | 8 | 745 | ||
| BC085706.1 | - | 8 | 869 | 7189102 | ||