siRNA Details
| Gene Symbol: Inpp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133562.1 | NP_598246.1 | 2 | 519 | ||
| XM_006251155.2 | XP_006251217.1 | 2 | 540 | |||
| XM_008770248.1 | XP_008768470.1 | 2 | 561 | |||
| GenBank mRNA | AB032551.1 | - | 2 | 519 | ||

Inpp5j
Genomic Map