siRNA Details
Gene Symbol: Giot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133563.1 | NP_598247.1 | 4 | 857 | ||
GenBank mRNA | AB047636.1 | - | 4 | 857 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Giot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133563.1 | NP_598247.1 | 4 | 857 | ||
GenBank mRNA | AB047636.1 | - | 4 | 857 |