siRNA Details
| Gene Symbol: Zbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133564.1 | NP_598248.1 | 7 | 1129 | ||
| XM_006235674.2 | XP_006235736.1 | 8 | 4972 | |||
| GenBank mRNA | AJ302054.1 | - | 7 | 1129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbp1| Gene Symbol: Zbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133564.1 | NP_598248.1 | 7 | 1129 | ||
| XM_006235674.2 | XP_006235736.1 | 8 | 4972 | |||
| GenBank mRNA | AJ302054.1 | - | 7 | 1129 | ||