siRNA Details
| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133565.1 | NP_598249.1 | 7 | 660 | ||
| GenBank mRNA | FQ217639.1 | - | 6 | 2661 | ||
| FQ217678.1 | - | 6 | 2660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G3bp1| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133565.1 | NP_598249.1 | 7 | 660 | ||
| GenBank mRNA | FQ217639.1 | - | 6 | 2661 | ||
| FQ217678.1 | - | 6 | 2660 | |||