siRNA Details
Gene Symbol: Angptl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133569.1 | NP_598253.1 | 4 | 1469 | ||
GenBank mRNA | AF159049.1 | - | 4 | 1469 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011923, AK214337, AK205240, AK037265, AK187902, and others.
Panther Classification:
