siRNA Details
| Gene Symbol: Grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133570.5 | NP_598254.2 | 2 | 324 | ||
| GenBank mRNA | BF567195.1 | - | 2 | 305 | ||
| M31176.1 | - | 2 | 728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grp| Gene Symbol: Grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133570.5 | NP_598254.2 | 2 | 324 | ||
| GenBank mRNA | BF567195.1 | - | 2 | 305 | ||
| M31176.1 | - | 2 | 728 | |||