siRNA Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133571.1 | NP_598255.1 | 10 | 1217 | ||
| XM_006243702.2 | XP_006243764.1 | 10 | 1439 | |||
| XM_006243703.1 | XP_006243765.1 | 11 | 990 | |||
| GenBank mRNA | D16236.1 | - | 10 | 1217 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007658, BC046296, AK077609, AK142334, AK077991, and others.

Cdc25a
Genomic Map