siRNA Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133572.1 | NP_598256.1 | 14 | 1671 | ||
| XM_006235003.1 | XP_006235065.1 | 13 | 1569 | |||
| GenBank mRNA | D16237.1 | - | 14 | 1671 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc25b| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133572.1 | NP_598256.1 | 14 | 1671 | ||
| XM_006235003.1 | XP_006235065.1 | 13 | 1569 | |||
| GenBank mRNA | D16237.1 | - | 14 | 1671 | ||