siRNA Details
| Gene Symbol: Gper1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133573.1 | NP_598257.1 | 1 | 502 | ||
| XM_006248914.2 | XP_006248976.1 | 2 | 1826 | |||
| GenBank mRNA | U92802.1 | - | 1 | 502 | ||
More Information
Other Species Targeted
Set Membership: |
|

Gper1
Genomic Map