siRNA Details
| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190993.1 | NP_001177922.1 | 7, 8 | 614 | ||
| XM_006257053.2 | XP_006257115.1 | 8, 9 | 646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arr3| Gene Symbol: Arr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190993.1 | NP_001177922.1 | 7, 8 | 614 | ||
| XM_006257053.2 | XP_006257115.1 | 8, 9 | 646 | |||