siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012745.1 | NP_001012763.1 | 2 | 675 | ||
| XM_006237187.2 | XP_006237249.1 | 2 | 667 | |||
| XM_006237189.2 | XP_006237251.1 | 2 | 495 | |||
| XM_006237190.2 | XP_006237252.1 | 2 | 526 | |||
| XM_008763249.1 | XP_008761471.1 | 2 | 603 | |||
| GenBank mRNA | AF281635.1 | - | 1 | 379 | ||
| AY724498.1 | - | 2 | 497 | |||
| BC090354.1 | - | 2 | 675 | 7319307 | ||

Zfp422
Genomic Map