siRNA Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133580.1 | NP_598264.1 | 7, 8 | 503 | ||
| GenBank mRNA | BC061984.1 | - | 8, 9 | 811 | 5599180 | |
| U18771.1 | - | 7, 8 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab26| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133580.1 | NP_598264.1 | 7, 8 | 503 | ||
| GenBank mRNA | BC061984.1 | - | 8, 9 | 811 | 5599180 | |
| U18771.1 | - | 7, 8 | 503 | |||