siRNA Details
| Gene Symbol: Blcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133582.3 | NP_598266.1 | 1 | 379 | ||
| GenBank mRNA | AB071598.1 | - | 1 | 347 | ||
| BC087661.1 | - | 1 | 172 | 7313852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blcap| Gene Symbol: Blcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133582.3 | NP_598266.1 | 1 | 379 | ||
| GenBank mRNA | AB071598.1 | - | 1 | 347 | ||
| BC087661.1 | - | 1 | 172 | 7313852 | ||