siRNA Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270862.1 | NP_001257791.1 | 12, 13 | 895 | ||
| NM_001270863.1 | NP_001257792.1 | 12, 13 | 877 | |||
| NM_001270864.1 | NP_001257793.1 | 11, 12 | 835 | |||
| NM_133583.2 | NP_598267.2 | 11, 12 | 853 | |||
| GenBank mRNA | AF334105.1 | - | 12, 13 | 859 | ||
| AF334106.1 | - | 11, 12 | 817 | |||
| AJ426424.1 | - | 12, 13 | 908 | |||
| AJ426425.1 | - | 11, 12 | 866 | |||
| AJ426426.1 | - | 12, 13 | 871 | |||
| AJ426427.1 | - | 11, 12 | 829 | |||
| FQ216465.1 | - | 4, 5 | 243 | |||

Ndrg2
Genomic Map