siRNA Details
| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133584.1 | NP_598268.1 | 12 | 2014 | ||
| XM_006233260.2 | XP_006233322.1 | 12 | 1942 | |||
| GenBank mRNA | D89093.1 | - | 12 | 2014 | ||
| U76032.1 | - | 3 | 130 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde5a| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133584.1 | NP_598268.1 | 12 | 2014 | ||
| XM_006233260.2 | XP_006233322.1 | 12 | 1942 | |||
| GenBank mRNA | D89093.1 | - | 12 | 2014 | ||
| U76032.1 | - | 3 | 130 | |||