siRNA Details
| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133584.1 | NP_598268.1 | 10, 11 | 1928 | ||
| XM_006233260.2 | XP_006233322.1 | 10, 11 | 1856 | |||
| GenBank mRNA | D89093.1 | - | 10, 11 | 1928 | ||
| U76032.1 | - | 1, 2 | 44 | |||

Pde5a
Genomic Map