siRNA Details
| Gene Symbol: Jmjd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191719.1 | NP_001178648.1 | 8 | 1836 | ||
| XM_006256359.2 | XP_006256421.1 | 8 | 1887 | |||
| XM_006256360.2 | XP_006256422.1 | 7 | 1683 | |||
| XM_006256361.2 | XP_006256423.1 | 8 | 1910 | |||
| XM_006256362.2 | XP_006256424.1 | 8 | 1802 | |||
| XM_006256363.1 | XP_006256425.1 | 4 | 1244 | |||
| XM_006256364.1 | XP_006256426.1 | 8 | 1796 | |||
| XM_006256365.1 | XP_006256427.1 | 3 | 1102 | |||

Jmjd1c
Genomic Map