siRNA Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133591.2 | NP_598275.1 | 6, 7 | 731 | ||
| XM_008767941.1 | XP_008766163.1 | 6, 7 | 1092 | |||
| XM_008767942.1 | XP_008766164.1 | 6, 7 | 1157 | |||
| XM_008767943.1 | XP_008766165.1 | 5, 6 | 1005 | |||
| GenBank mRNA | AF022774.1 | - | 5, 6 | 657 | ||
| BC091126.1 | - | 6, 7 | 731 | 7380477 | ||

Rph3al
Genomic Map