siRNA Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133593.2 | NP_598277.2 | 7 | 1030 | ||
| XM_006251629.1 | XP_006251691.1 | 7 | 1557 | |||
| XM_006251630.2 | XP_006251692.1 | 7 | 1076 | |||
| GenBank mRNA | BC070925.1 | - | 7 | 1030 | 7096586 | |
| L07073.1 | - | 7 | 1043 | |||

Ap3m1
Genomic Map