siRNA Details
| Gene Symbol: Cblb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133601.1 | NP_598285.1 | 3 | 309 | ||
| XM_006248258.2 | XP_006248320.1 | 3 | 383 | |||
| XM_006248259.2 | XP_006248321.1 | 3 | 466 | |||
| XM_006248260.2 | XP_006248322.1 | 3 | 382 | |||
| XM_008768626.1 | XP_008766848.1 | 2 | 284 | |||
| XM_008768627.1 | XP_008766849.1 | 3 | 579 | |||
| GenBank mRNA | AB071283.1 | - | 3 | 309 | ||

Cblb
Genomic Map