siRNA Details
| Gene Symbol: Ehhadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133606.1 | NP_598290.1 | 7 | 1752 | ||
| GenBank mRNA | BC089777.1 | - | 7 | 1741 | 7321434 | |
| K03249.1 | - | 7 | 1752 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehhadh| Gene Symbol: Ehhadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133606.1 | NP_598290.1 | 7 | 1752 | ||
| GenBank mRNA | BC089777.1 | - | 7 | 1741 | 7321434 | |
| K03249.1 | - | 7 | 1752 | |||