siRNA Details
Gene Symbol: Trpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136229.1 | NP_001129701.1 | 11 | 1503 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136229.1 | NP_001129701.1 | 11 | 1503 |