siRNA Details
| Gene Symbol: Trpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136229.1 | NP_001129701.1 | 14 | 1941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpm4| Gene Symbol: Trpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136229.1 | NP_001129701.1 | 14 | 1941 | ||