siRNA Details
| Gene Symbol: Slco4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133608.2 | NP_598292.2 | 11 | 2141 | ||
| GenBank mRNA | AF239262.1 | - | 10 | 1975 | ||
| BC097285.1 | - | 11 | 2141 | 7441748 | ||
| BC097286.1 | - | 11 | 2083 | 7444297 | ||

Slco4a1
Genomic Map