siRNA Details
| Gene Symbol: Eif2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133609.2 | NP_598293.2 | 8 | 1034 | ||
| XM_006238630.2 | XP_006238692.1 | 8 | 1005 | |||
| GenBank mRNA | BC072507.1 | - | 8 | 1034 | 7099973 | |
| U38253.1 | - | 8 | 1014 | |||

Eif2b3
Genomic Map