siRNA Details
| Gene Symbol: Serpini2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134409.1 | NP_001127881.1 | 1 | 79 | ||
| XM_008761077.1 | XP_008759299.1 | 2 | 252 | |||
| GenBank mRNA | X99773.1 | - | 1 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpini2| Gene Symbol: Serpini2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134409.1 | NP_001127881.1 | 1 | 79 | ||
| XM_008761077.1 | XP_008759299.1 | 2 | 252 | |||
| GenBank mRNA | X99773.1 | - | 1 | 94 | ||