siRNA Details
| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133614.2 | NP_598298.1 | 6 | 606 | ||
| XM_006240114.2 | XP_006240176.1 | 6 | 1932 | |||
| GenBank mRNA | AJ289714.1 | - | 6 | 450 | ||
| BC089099.1 | - | 6 | 606 | 7306738 | ||

Slc25a21
Genomic Map