siRNA Details
| Gene Symbol: Zhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133620.1 | NP_598304.1 | 3 | 2520 | ||
| GenBank mRNA | AB072439.1 | - | 3 | 2520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zhx1| Gene Symbol: Zhx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133620.1 | NP_598304.1 | 3 | 2520 | ||
| GenBank mRNA | AB072439.1 | - | 3 | 2520 | ||