siRNA Details
| Gene Symbol: Reg3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145846.2 | NP_001139318.1 | 3 | 217 | ||
| NM_172077.2 | NP_742074.2 | 3 | 319 | |||
| GenBank mRNA | CF249035.1 | - | 2 | 206 | ||
| D23676.1 | - | 3 | 217 | |||
| L10229.1 | - | 3 | 319 | |||

Reg3a
Genomic Map