siRNA Details
| Gene Symbol: Reg3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145846.2 | NP_001139318.1 | 5 | 502 | ||
| NM_172077.2 | NP_742074.2 | 5 | 604 | |||
| GenBank mRNA | D23676.1 | - | 5 | 502 | ||
| L10229.1 | - | 5 | 604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reg3a| Gene Symbol: Reg3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145846.2 | NP_001139318.1 | 5 | 502 | ||
| NM_172077.2 | NP_742074.2 | 5 | 604 | |||
| GenBank mRNA | D23676.1 | - | 5 | 502 | ||
| L10229.1 | - | 5 | 604 | |||