siRNA Details
| Gene Symbol: Ctsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134334.2 | NP_599161.2 | 2 | 318 | ||
| GenBank mRNA | BC062032.1 | - | 2 | 318 | 5623627 | |
| X54467.1 | - | 2 | 263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsd| Gene Symbol: Ctsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134334.2 | NP_599161.2 | 2 | 318 | ||
| GenBank mRNA | BC062032.1 | - | 2 | 318 | 5623627 | |
| X54467.1 | - | 2 | 263 | |||