siRNA Details
| Gene Symbol: Myo9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134335.1 | NP_599162.1 | 6 | 1553 | ||
| GenBank mRNA | AJ001713.1 | - | 6 | 1553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo9a| Gene Symbol: Myo9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134335.1 | NP_599162.1 | 6 | 1553 | ||
| GenBank mRNA | AJ001713.1 | - | 6 | 1553 | ||