siRNA Details
| Gene Symbol: Kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169112.1 | NP_001162583.1 | 5 | 676 | ||
| GenBank mRNA | AF035955.1 | - | 2 | 184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif11| Gene Symbol: Kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169112.1 | NP_001162583.1 | 5 | 676 | ||
| GenBank mRNA | AF035955.1 | - | 2 | 184 | ||