siRNA Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170604.1 | NP_001164075.1 | 11 | 1305 | ||
| XM_006252821.2 | XP_006252883.1 | 12 | 1401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12rb1| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170604.1 | NP_001164075.1 | 11 | 1305 | ||
| XM_006252821.2 | XP_006252883.1 | 12 | 1401 | |||