siRNA Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191750.1 | NP_001178679.1 | 13 | 1968 | ||
| XM_006236613.2 | XP_006236675.1 | 15 | 2362 | |||
| XM_006236614.2 | XP_006236676.1 | 15 | 2358 | |||
| GenBank mRNA | AF083329.1 | - | 2 | 122 | ||

Il12rb2
Genomic Map