siRNA Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318230.1 | NP_001305159.1 | 3 | 711 | ||
| NM_006285.2 | NP_006276.2 | 3 | 711 | |||
| GenBank mRNA | AF507907.1 | - | 2 | 124 | ||
| AK298907.1 | - | 3 | 249 | |||
| BC037561.1 | - | 3 | 306 | |||
| BC038448.1 | - | 3 | 685 | 5170888 | ||
| BC067130.1 | - | 3 | 722 | 6153652 | ||
| D50863.1 | - | 3 | 647 | |||

TESK1
Genomic Map