siRNA Details
| Gene Symbol: Eef1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175838.1 | NP_787032.1 | 7, 8 | 1337 | ||
| GenBank mRNA | BC063162.1 | - | 7, 8 | 1333 | 6918979 | |
| BC072542.1 | - | 7, 8 | 1332 | 7103726 | ||
| BC091297.1 | - | 7, 8 | 1318 | 7309009 | ||
| BC111707.1 | - | 6, 7 | 1301 | 7367097 | ||
| BC128723.1 | - | 7, 8 | 1312 | 7312569 | ||
| L10339.1 | - | 6, 7 | 1272 | |||
| M81088.1 | - | 2, 3 | 498 | |||
| X61043.1 | - | 7, 8 | 1337 | |||
| X63561.1 | - | 6, 7 | 1310 | |||

Eef1a1
Genomic Map