siRNA Details
| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134359.1 | NP_599186.1 | 4, 5 | 483 | ||
| XM_006230206.1 | XP_006230268.1 | 5, 6 | 479 | |||
| GenBank mRNA | BC091574.1 | - | 4, 5 | 483 | 7135215 | |
| FQ216602.1 | - | 5, 6 | 439 | |||

Ppp4c
Genomic Map