siRNA Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134364.1 | NP_599191.1 | 2 | 262 | ||
| GenBank mRNA | BC099743.1 | - | 2 | 245 | 7937989 | |
| M19044.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5b| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134364.1 | NP_599191.1 | 2 | 262 | ||
| GenBank mRNA | BC099743.1 | - | 2 | 245 | 7937989 | |
| M19044.1 | - | 1 | 78 | |||