siRNA Details
| Gene Symbol: Smarca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134368.1 | NP_599195.1 | 20, 21 | 3303 | ||
| XM_006242596.2 | XP_006242658.1 | 20, 21 | 3300 | |||
| XM_006242597.2 | XP_006242659.1 | 20, 21 | 3300 | |||
| XM_006242598.2 | XP_006242660.1 | 20, 21 | 3300 | |||
| XM_006242599.2 | XP_006242661.1 | 20, 21 | 3300 | |||
| XM_006242600.2 | XP_006242662.1 | 20, 21 | 3300 | |||
| XM_006242601.2 | XP_006242663.1 | 20, 21 | 3300 | |||
| GenBank mRNA | AJ504723.1 | - | 19, 20 | 2981 | ||

Smarca4
Genomic Map